MilliporeSigma
  • Phosphotriesterase: a complementary tool for the selective detection of two organophosphate insecticides: chlorpyrifos and chlorfenvinfos.
Phosphotriesterase: a complementary tool for the selective detection of two organophosphate insecticides: chlorpyrifos and chlorfenvinfos.
Talanta (2009-01-23)
Georges Istamboulie, Didier Fournier, Jean-Louis Marty, Thierry Noguer
ABSTRACT
This work shows the possibility of combining the high sensitivity of genetically-modified Drosophila melanogaster acetylcholinesterase (B394) with the ability of phosphotriesterase (PTE) to hydrolyse organophosphate compounds, in the aim of developing a biosensor selective to two insecticides of interest: chlorpyrifos and chlorfenvinfos. The studies clearly demonstrate that chlorfenvinfos is a substrate that acts as competitive inhibitor of PTE, therefore preventing the efficient hydrolysis of other pesticides, including chlorpyrifos. A bi-enzymatic sensor was designed by immobilizing both B394 and PTE in a polyvinylalcohol matrix. The sensor was shown to be able to discriminate between chlorpyrifos and chlorfenvinfos inhibitions.
Product Description
Supelco
Chlorfenvinphos, PESTANAL®, analytical standard