MilliporeSigma
  • A novel administration route for edaravone: I. Effects of metabolic inhibitors on skin permeability of edaravone.
A novel administration route for edaravone: I. Effects of metabolic inhibitors on skin permeability of edaravone.
International journal of pharmaceutics (2009-01-27)
Toshiaki Sato, Keizo Mizuno, Fumiyoshi Ishii
ABSTRACT
We examined the effects of metabolic inhibitors on skin permeation of edaravone. SKF-525A, diclofenac sodium (DIC) and indomethacin (IND) were added to supernatant fluid (SF) of hairless rat (HR) skin homogenate. L-Cysteine (L-Cys) and benzotriazole (BTA), as pharmaceutical additives, were added to HR skin homogenate SF, and incubated at 37 degrees C for 30 min. K(m) and V(max) values were calculated. For determination of edaravone skin permeation from edaravone/hydroxypropyl-beta-cyclodextrin (HPbetaCD) complex solution, HR skin was placed in a Franz diffusion cell, and kept at 37 degrees C. Edaravone/HPbetaCD solution that contained L-Cys was put into the donor side. The relative activity in skin homogenate SF after co-treatment with IND and SKF-525A decreased to 40.8% of the control. However, DIC and IND had a weak inhibitory effect. For inhibition of edaravone metabolism, L-Cys and BTA had no effect on K(m) value, but V(max) was significantly decreased compared with controls (*P<0.05, Tukey-Kramer test). The edaravone skin permeation rate and permeability coefficient from edaravone/HPbetaCD complex solution with inhibitor were significantly increased compared with those without inhibitor. We suggest that the metabolism inhibitor was useful for the transdermal delivery of edaravone.
Product Description
Supelco
Proadifen hydrochloride, analytical standard, ≥95%