MilliporeSigma
  • Detection of enzymatically generated hydrogen peroxide by metal-based fluorescent probe.
Detection of enzymatically generated hydrogen peroxide by metal-based fluorescent probe.
Analytical chemistry (2011-11-18)
Yutaka Hitomi, Toshiyuki Takeyasu, Takuzo Funabiki, Masahito Kodera
ABSTRACT
We developed a metal-based fluorescent probe for H(2)O(2) called MBFh1, which has an iron complex as a reaction site for H(2)O(2) and a 3,7-dihydroxyphenoxazine derivative as the fluorescent reporter unit. The iron complex reacts quickly with H(2)O(2) to form oxidants, and then the oxidants convert the closely appended nonfluorescent 3,7-dihydroxyphenoxazine moiety to resorufin in an intramolecular fashion. The quick response to H(2)O(2) allows us to plot the enzymatic evolution of H(2)O(2). A combination of N-acetyl-3,7-dihydroxyphenoxazine and horseradish peroxidase has been frequently used to detect enzymatically generated H(2)O(2), but this method has interference with phenol derivatives. The use of MBFh1 overcomes this drawback.