MilliporeSigma
  • First kinetic evidence for the CH/π and π/π solute-solvent interaction of C60 in the Diels-Alder reaction with cyclohexadiene.
First kinetic evidence for the CH/π and π/π solute-solvent interaction of C60 in the Diels-Alder reaction with cyclohexadiene.
Organic & biomolecular chemistry (2012-01-13)
Takumi Oshima, Tsubasa Mikie, Naohiko Ikuma, Hajime Yakuma
ABSTRACT
The first CH/π solute-solvent interaction of C(60) was evidenced by the kinetic solvent effects in the Diels-Alder reaction with 1,3-cyclohexadiene based on the evaluation of linear free energy relationship of log k(2) with empirical solvent polarity and basicity parameters, E(T)(30) and D(π), respectively.
Sigma-Aldrich
1,3-Cyclohexadiene, contains 0.05% BHT as inhibitor, 97%