MilliporeSigma
  • Specific interaction of the new fluorescent dye 10-N-nonyl acridine orange with inner mitochondrial membrane. A lipid-mediated inhibition of oxidative phosphorylation.
Specific interaction of the new fluorescent dye 10-N-nonyl acridine orange with inner mitochondrial membrane. A lipid-mediated inhibition of oxidative phosphorylation.
FEBS letters (1990-01-29)
A Maftah, J M Petit, R Julien
ABSTRACT
The fluorescent dye 10-N-nonyl acridine orange (NAO), known as specifically associated with mitochondria, has been reported to have a cytotoxic effect when high doses were applied to cells. Presently, the biochemical basis of its toxicity was investigated on isolated rat liver mitochondria. At low concentrations, NAO strongly inhibited state 3 respiration and ATP synthesis. At high concentrations, electron transport, ATP hydrolysis, Pi-transport and adenine nucleotide activities were also decreased. All these inhibitions can be explained by probe-cardiolipin interactions which could induce the collapse of energy conversion and/or the modification of membrane fluidity.