MilliporeSigma
  • Effects of 5 beta-chol-3-en-24-oic acid, and lithocholic acid and its sulfates on prostaglandin E2 output in perfusion of the rat colon.
Effects of 5 beta-chol-3-en-24-oic acid, and lithocholic acid and its sulfates on prostaglandin E2 output in perfusion of the rat colon.
Gastroenterologia Japonica (1989-02-01)
Y Hikasa, N Tanida, K Sawada, K Furukawa, M Kano, T Shimoyama
ABSTRACT
The effects of bile salts on the output of prostaglandin E2(PGE2) were studied in rats by colonic perfusion. Bile salts and their concentrations in infusates were as follows; chenodeoxycholic acid sodium salt (CDCNa), sulfolithocholic acid disodium salt (SLCNa) and sulfotaurolithocholic acid disodium salt (STLCNa) were at 1 mmol/l, lithocholic acid sodium salt (LCNa) was at 0.6 mmol/l, 5 beta-chol-3-en-24-oic acid sodium salt (delta 3Na) was at 0.006 mmol/l, and sulfoglycolithocholic acid disodium salt (SGLCNa) was at 0.8 mmol/l. Median values of PGE2 outputs were 40.9, 52.4, 65.5, trace, 7.6, 9.8 and 9.8 pg/10 min/cm in the CDCNa, LCNa, delta 3Na, SLCNa, STLCNa, SGLCNa and control groups, respectively (control vs CDCNa, LCNa, delta 3Na, SLCNa group, p less than 0.01 by median test). There was such a dissociation in PGE2 output and electrolyte concentration in the perfusates that high concentration of Na+ and Cl- ions was observed only in the SGLCNa group (p less than 0.01 by t-test compared with the control group), whereas in the remainder of the groups substantial net movement of electrolytes and water was not observed. Light microscopy showed no evidence of morphological damage in any group. These results indicated that CDCNa, LCNa and delta 3Na increased PGE2 outputs in the colon at low concentration without functional or morphological changes.