MilliporeSigma
  • Renal tubular excretion of the N4-acetyl metabolites of sulphasomidine and sulphadimethoxine in the dog.
Renal tubular excretion of the N4-acetyl metabolites of sulphasomidine and sulphadimethoxine in the dog.
The Journal of pharmacy and pharmacology (1993-07-01)
S P Boom, A C Wouterse, T B Vree, C A Van Ginneken, F G Russel
ABSTRACT
To investigate whether dogs are able to excrete acetylated drugs by active transport, the plasma kinetics and renal excretion of the N4-acetyl metabolites of sulphasomidine and sulphadimethoxine were studied in the beagle dog after a rapid intravenous bolus injection. Two doses of N4-acetylsulphasomidine (1050 and 105 mg) and one dose of N4-acetylsulphadimethoxine (472 mg) were administered on separate occasions. The renal clearance (CLR) was as follows: N4-acetylsulphasomidine (1050 mg) 34 mL min-1; N4-acetylsulphasomidine (105 mg) 28 mL min-1; and N4-acetylsulphadimethoxine (472 mg) 24 mL min-1. CLR was higher than expected on the basis of the measured glomerular filtration rate, indicating that the N4-acetyl metabolites may be excreted by the renal tubules by active tubular transport. Saturation of the excretion process of N4-acetylsulphasomidine occurred with a transport maximum of 930 +/- 190 micrograms min-1 and a Michaelis-Menten constant of 37 +/- 10 micrograms mL-1. It may be concluded that the dog renal organic anion transport system is able to secrete acetylated sulphonamides.