MilliporeSigma
  • In situ characterization of nonmitochondrial Ca2+ stores in individual pancreatic beta-cells.
In situ characterization of nonmitochondrial Ca2+ stores in individual pancreatic beta-cells.
Diabetes (1998-08-14)
A Tengholm, C Hagman, E Gylfe, B Hellman
ABSTRACT
Free Ca2+ was measured in intracellular stores of individual mouse pancreatic beta-cells using dual-wavelength microfluorometry and the low-affinity Ca2+ indicator furaptra. Controlled permeabilization of the plasma membrane with 4 micromol/l digitonin revealed that 22% of the furaptra was trapped in intracellular nonnuclear compartments. When 3 mmol/l ATP and 200 nmol/l Ca2+ were simultaneously present, this cation rapidly accumulated in the organelle pool, reaching an average concentration of 200-500 micromol/l. Whereas agents affecting the mitochondrial function (5 mmol/l succinate, 2 micromol/l ruthenium red, or 10 micromol/l antimycin A + 2 microg/ml oligomycin) had little effects, the Ca2+-ATPase inhibitor thapsigargin released 92% of the Ca2+ mobilizable with the ionophore Br-A23187. Digital imaging revealed regional differences in the organelle Ca2+. The regions with the highest Ca2+ concentration were particularly responsive to inositol 1,4,5-trisphosphate (IP3). IP3 mobilized Ca2+ in a dose-dependent way with half-maximal and maximal effects at about 1 and 5 micromol/l, respectively. High concentrations of IP3 released about half of the thapsigargin-sensitive Ca2+, but there were no responses to agents known to activate ryanodine receptors, such as 10 mmol/l caffeine, 0.1-1 micromol/l ryanodine, or 1-5 micromol/l cyclic ADP ribose. The results reinforce the concept that mobilization of intracellular Ca2+ in the pancreatic beta-cell is mediated by IP3 receptors rather than ryanodine receptors.
Sigma-Aldrich
4-Bromo-calcium Ionophore A23187, powder