MilliporeSigma
  • Murine ortholog of ASC, a CARD-containing protein, self-associates and exhibits restricted distribution in developing mouse embryos.
Murine ortholog of ASC, a CARD-containing protein, self-associates and exhibits restricted distribution in developing mouse embryos.
Experimental cell research (2001-01-05)
J Masumoto, S Taniguchi, K Nakayama, K Ayukawa, J Sagara
ABSTRACT
ASC (apoptosis-associated speck-like protein containing a CARD) was first identified as a cytosolic soluble protein that forms insoluble aggregates and enhances etoposide-induced apoptosis. We have cloned a murine ortholog of ASC (mASC) comprising 193 amino acids with a well-conserved pyrin N-terminal homology domain and caspase recruitment domain (CARD). mASC fused with green fluorescent protein appeared as a speck in transfected COS-7 cells and showed self-association. We analyzed mASC gene expression in developing embryos by in situ hybridization and found it to have a restricted distribution in mouse embryos. At E9.5, mASC was strongly expressed in the telencephalon, thalamic areas of the diencephalon, heart, and liver. Northern blotting analysis revealed that the mASC gene was expressed ubiquitously in multiple organs in adult mice. These findings indicate that mASC shows conservation of not only the primary structure of human ASC but also the ability to aggregate and has some similarity in its distribution to other CARD-containing molecules, including the apoptosis regulator Apaf-1.