MilliporeSigma
  • An arginine residue is essential for stretching and binding of the substrate on UDP-glucose-4-epimerase from Escherichia coli. Use of a stacked and quenched uridine nucleotide fluorophore as probe.
An arginine residue is essential for stretching and binding of the substrate on UDP-glucose-4-epimerase from Escherichia coli. Use of a stacked and quenched uridine nucleotide fluorophore as probe.
The Journal of biological chemistry (1999-05-18)
U Bhattacharyya, G Dhar, A Bhaduri
ABSTRACT
In the previous paper we demonstrated that uridine-5'-beta-1-(5-sulfonic acid) naphthylamidate (UDPAmNS) is a stacked and quenched fluorophore that shows severalfold enhancement of fluorescence in a stretched conformation. UDPAmNS was found to be a powerful competitive inhibitor (Ki = 0.2 mM) for UDP-glucose-4-epimerase from Escherichia coli. This active site-directed fluorophore assumed a stretched conformation on the enzyme surface, as was evidenced by full enhancement of fluorescence in saturating enzyme concentration. Complete displacement of the fluorophore by UDP suggested it to bind to the substrate binding site of the active site. Analysis of inactivation kinetics in presence of alpha,beta-diones such as phenylglyoxal, cyclohaxanedione, and 2,3-butadione suggested involvement of the essential arginine residue in the overall catalytic process. From spectral analysis, loss of activity could also be directly correlated with modification of only one arginine residue. Protection experiments with UDP showed the arginine residue to be located in the uridyl phosphate binding subsite. Unlike the native enzyme, the modified enzyme failed to show any enhancement of fluorescence with UDPAmNS clearly demonstrating the role of the essential arginine residue in stretching and binding of the substrate. The potential usefulness of such stacked and quenched nucleotide fluorophores has been discussed.