MilliporeSigma
  • Analogs of arginine vasopressin modified in position 3 with (R)-alpha-hydroxymethylphenylalanine.
Analogs of arginine vasopressin modified in position 3 with (R)-alpha-hydroxymethylphenylalanine.
The journal of peptide research : official journal of the American Peptide Society (1999-07-29)
E Lempicka, J Slaninova, A Olma, B Lammek
ABSTRACT
This study describes the synthesis and some pharmacological properties of three new analogs of arginine vasopressin (AVP) substituted in position 3 with (R)-alpha-hydroxymethylphenylalanine ([R]-HmPhe). All new peptides were tested for vasopressor and antidiuretic as well as uterotonic activity. None of the 3 analogs showed any pressor activity and their uterotonic activity was negligible. Only analog [Mpa1,(R)-HmPhe3]AVP exhibited significant antidiuretic activity.