MilliporeSigma
  • Reinvestigation of the reaction between 2-furancarboxaldehyde and 4-hydroxy-5-methyl-3(2H)-furanone.
Reinvestigation of the reaction between 2-furancarboxaldehyde and 4-hydroxy-5-methyl-3(2H)-furanone.
Journal of agricultural and food chemistry (1999-12-22)
A Ravagli, G Boschin, L Scaglioni, A Arnoldi
ABSTRACT
The reaction between 2-furancarboxaldehyde and 4-hydroxy-5-methyl-3(2H)-furanone was reinvestigated as a part of a systematic study on low molecular weight colored compounds from the Maillard reaction. In acetic acid/piperidine, besides 2-(2-furanylmethylene)-4-hydroxy-5-methyl-3(2H)-furanone (1) and 5-[2-(2-furanyl)ethenyl]-2-(2-furanylmethylene)-4-hydroxy-5-methyl -3( 2H)-furanone (2), four novel compounds, 15a, 15b, 16a, and 16b, were isolated and characterized. These compounds are produced from two molecules of furanone 1 and one molecule of 2-furancarboxaldehyde, and a mechanism is proposed for their formation. Compounds 1, 15a, 15b, 16a, and 16b are formed also by reacting 2-furancarboxaldehyde and 4-hydroxy-5-methyl-3(2H)-furanone in water at pH 3 and 2, whereas 2 was never detected. The formation of these compounds was studied also in xylose/lysine and xylose/glycine model systems.
Sigma-Aldrich
4-Hydroxy-5-methyl-3-furanone, 97%, FG
Sigma-Aldrich
4-Hydroxy-5-methyl-3-furanone, 97%