MilliporeSigma
  • Formaldehyde activation of mitoxantrone yields CpG and CpA specific DNA adducts.
Formaldehyde activation of mitoxantrone yields CpG and CpA specific DNA adducts.
Nucleic acids research (2000-01-29)
B S Parker, S M Cutts, C Cullinane, D R Phillips
ABSTRACT
Recently we have found that mitoxantrone, like Adria-mycin, can be activated by formaldehyde and subsequently form adducts which stabilise double-stranded DNA in vitro. This activation by formaldehyde may be biologically relevant since formaldehyde levels are elevated in those tumours in which mitoxan-trone is most cytotoxic. In vitro transcription analysis revealed that these adducts block the progression of RNA polymerase during transcription and cause truncated RNA transcripts. There was an absolute requirement for both mitoxantrone and formaldehyde in transcriptional blockage formation and the activated complex was found to exhibit site specificity, with blockage occurring prior to CpG and CpA sites in the DNA (non-template strand). The stability of the adduct at 37 degrees C was site dependent. The half-lives ranged from 45 min to approximately 5 h and this was dependent on both the central 2 bp blockage site as well as flanking sequences. The CpG specificity of mitoxantrone adduct sites was also confirmed independently by a lambda exonuclease digestion assay.