MilliporeSigma
  • A simplified procedure for purification of cytochrome P-450 by preparative ampholine gel for isoelectric focusing.
A simplified procedure for purification of cytochrome P-450 by preparative ampholine gel for isoelectric focusing.
Preparative biochemistry & biotechnology (2000-05-04)
M Kuwada, J Maki, H Hasumi, S Furudate, K Takahasi
ABSTRACT
The purification process for cytochrome P450 is very complicated, involving five or more column chromatography steps for the final preparation. This paper describes a reduction in the number of the steps; it can be easily purified from pig testis microsomes with improved the yield. As the first step, DEAE-Toyopearl column chromatography is performed only once and then, as the second step, the partially purified cytochrome P450 is completely purified by a preparative Ampholine PAG-plate Gel for Isoelectric Focusing. The combination reduced the purification to a two-step procedure.