MilliporeSigma
  • [The use of nitatsid and gipozol'-N in the combined treatment of acute inflammatory diseases of the maxillofacial area].
[The use of nitatsid and gipozol'-N in the combined treatment of acute inflammatory diseases of the maxillofacial area].
Stomatologiia (2000-05-17)
S V Tarasenko, I P Aleksashina, V S Agapov, G M Trukhina, N A Liapunov
ABSTRACT
Combined drugs with a hydrophilic base, nitacide and hyposol-n, were used in combined therapy of acute purulent inflammations of the maxillofacial area. Time course of clinical parameters and changes in microflora were studied in patients administered different local treatments with different inflammatory reactions. The data indicate a high efficiency of the drugs, which optimized local therapy of suppurative wounds and are recommended for wide practical use.