MilliporeSigma
  • In vitro inhibitory effect of rupatadine on histamine and TNF-alpha release from dispersed canine skin mast cells and the human mast cell line HMC-1.
In vitro inhibitory effect of rupatadine on histamine and TNF-alpha release from dispersed canine skin mast cells and the human mast cell line HMC-1.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2000-08-26)
M Queralt, P Brazís, M Merlos, F de Mora, A Puigdemont
ABSTRACT
To examine the inhibitory potential of rupatadine, a new H1-antihistamine and anti-PAF agent, on histamine and TNF-alpha release. Comparison with an H1-antihistamine (loratadine) and a PAF-antagonist (SR-27417A). Dispersed canine skin mast cells were used to assess the effect of the drugs tested on FcepsilonRI-dependent and -independent histamine release; the human HMC-1 cell line was used to study TNF-alpha release. Before stimulation mast cell populations were treated with increasing concentrations of rupatadine, loratadine and SR-27417A. Histamine and TNF-alpha release were measured following 15-30 min and 3 h activation, respectively. The IC50 for rupatadine in A23187, concanavalin A and anti-IgE induced histamine release was 0.7+/-0.4 microM, 3.2+/-0.7 microM and 1.5+/-0.4 microM, respectively whereas for loratadine the IC50 was 2.1+/-0.9 microM, 4.0+/-1.3 M and 1.7+/-0.5 microM. SR-27417A exhibited no inhibitory effect. Rupatadine, loratadine and SR-27417A inhibited TNF-alpha release with IC50 2.0+/-0.9 microM, 2.1+/-1.1 M and 4.3+/-0.6 microM, respectively. Rupatadine and loratadine showed similar inhibitory effect on histamine and TNF-alpha release, whereas SR-27417A only exhibited inhibitory effect against TNF-alpha.
Sigma-Aldrich
Rupatadine fumarate, ≥98% (HPLC)
Sigma-Aldrich
SR 27417, ≥98% (HPLC)