MilliporeSigma
  • Self-augmentation effect of male-specific products on sexually differentiated progesterone metabolism in adult male rat liver microsomes.
Self-augmentation effect of male-specific products on sexually differentiated progesterone metabolism in adult male rat liver microsomes.
The Journal of biological chemistry (2000-09-21)
A Yamada, M Yamada, Y Fujita, T Nishigami, K Nakasho, K Uematsu
ABSTRACT
It is well known that several 3-keto-4-ene steroids such as progesterone and testosterone are metabolized in a gender-specific or -predominant manner by adult rat liver microsomes. In the male, these steroids are primarily metabolized into two oxidized (16alpha-hydroxyl and 6beta-hydroxyl) products mainly by the respective, male-specific cytochrome P450 subforms, CYP2C11 and CYP3A2, while they are primarily metabolized into the 5alpha-reduced products by female-predominant 5alpha-reductase in the female. These sexually differentiated enzyme activities are largely regulated at the transcription level under endocrine control. In the present study, we show that unlabeled 16alpha-hydroxyprogesterone and 6beta-hydroxyprogesterone inhibited the 5alpha-reductive [(3)H]progesterone metabolism by adult male rat liver microsomes without significantly inhibiting the CYP2C11 and CYP3A2 activities producing themselves, whereas 3alpha-hydroxy-5alpha-pregnan-20-one and 5alpha-pregnane-3,20-dione not only stimulated the 5alpha-reductive metabolism producing themselves but also inhibited the male-specific oxidative metabolism. This finding compels us to propose a novel hypothesis that adult male rat liver microsomes may possess a self-augmentation system regulated by the male-specific products on sexually differentiated steroid metabolism, besides regulation by gene expressions of the related enzymes.