MilliporeSigma
  • Synthesis of 6-acetamido-5-amino- and -5-guanidino-3, 4-dehydro-N-(2-ethylbutyryl)- 3-piperidinecarboxylic acids related to zanamivir and oseltamivir, inhibitors of influenza virus neuraminidases.
Synthesis of 6-acetamido-5-amino- and -5-guanidino-3, 4-dehydro-N-(2-ethylbutyryl)- 3-piperidinecarboxylic acids related to zanamivir and oseltamivir, inhibitors of influenza virus neuraminidases.
Organic letters (2000-12-02)
E Shitara, Y Nishimura, K Nerome, Y Hiramoto, T Takeuchi
ABSTRACT
[reaction: see text] 6-Acetamido-5-amino- and -5-guanidino-3, 4-dehydro-N-(2-ethylbutyryl)-3-piperidinecarboxylic acids (8 and 9) have been synthesized starting from natural siastatin B, a bacterial neuraminidase inhibitor isolated from Streptomyces culture in a stereospecific fashion. These compounds are related to zanamivir and oseltamivir, inhibitors of influenza virus neuraminidases.
Sigma-Aldrich
Siastatin B, lyophilized powder, from microbial