MilliporeSigma
  • Chirality organization of ferrocenes bearing podand dipeptide chains: synthesis and structural characterization.
Chirality organization of ferrocenes bearing podand dipeptide chains: synthesis and structural characterization.
Journal of the American Chemical Society (2001-03-29)
T Moriuchi, A Nomoto, K Yoshida, A Ogawa, T Hirao
ABSTRACT
A variety of ferrocenes bearing podand dipeptide chains have been synthesized to form an ordered structure in both solid and solution states and have been investigated by 1H NMR, FT-IR, CD, and X-ray crystallographic analyses. Conformational enantiomerization through chirality organization was achieved by the intramolecular hydrogen bondings between the podand dipeptide chains. The single-crystal X-ray structure determination of the ferrocene 2 bearing the podand dipeptide chains (-D-Ala-D-Pro-OEt) revealed two C2-symmetric intramolecular hydrogen bondings between CO (Ala) and NH (another Ala) of each podand dipeptide chain to induce the chirality-organized structure. The molecular structures of the ferrocene 1 composed of the podand L-dipeptide chains (-L-Ala-L-Pro-OEt) and 2 are in a good mirror image relationship, indicating that they are conformational enantiomers. An opposite helically ordered molecular arrangement was formed in the crystal packing of 2 as compared with 1. The ferrocene 2 exhibited induced circular dichroism (CD), which appeared at the absorbance of the ferrocene moiety. The mirror image of the CD signals between 1 and 2 was observed, suggesting that the chirality-organized structure via intramolecular hydrogen bondings is present even in solution. The ferrocene 4 bearing the podand dipeptide chains (-Gly-L-Leu-OEt) also showed an ordered structure in the crystal based on two intramolecular hydrogen bondings between CO (Gly) and NH (another Gly) of each podand dipeptide chain, together with intermolecular hydrogen bondings between CO adjacent to the ferrocene unit and NH (neighboring Leu) to create the highly organized self-assembly. A different self-assembly was observed in the crystal of the ferrocene 5 composed of the podand dipeptide chains (-Gly-L-Phe-OEt), wherein each molecule is bonded to two neighboring molecules through two pairs of symmetrical intermolecular hydrogen bonds to form a 14-membered intermolecularly hydrogen-bonded ring. These ordered structures based on the intramolecular hydrogen bondings in the solution state are also confirmed by 1H NMR and FT-IR.
Sigma-Aldrich
Ala-Pro hydrate