MilliporeSigma
  • Synthesis of branched cyclomaltooligosaccharide carboxylic acids (cyclodextrin carboxylic acids) by microbial oxidation.
Synthesis of branched cyclomaltooligosaccharide carboxylic acids (cyclodextrin carboxylic acids) by microbial oxidation.
Carbohydrate research (2001-06-12)
T Ishiguro, T Fuse, M Oka, T Kurasawa, M Nakamichi, Y Yasumura, M Tsuda, T Yamaguchi, I Nogami
ABSTRACT
Novel branched cyclomaltooligosaccharide carboxylic acid (cyclodextrin carboxylic acid) derivatives were synthesized by microbial oxidation using Pseudogluconobacter saccharoketogenes to oxidize five types of branched cyclodextrins, including maltosyl beta-cyclodextrin (maltosyl-beta-CyD). For each novel cyclodextrin carboxylic acid derivative synthesized, the hydroxymethyl group of the terminal glucose residue in the branched part of the molecule was regiospecifically oxidized to a carboxyl group to give the corresponding uronic acid. In addition, the physicochemical properties of cyclomaltoheptaosyl-(6-->1)-alpha-D-glucopyranosyl-(4-->1)-alpha-D-glucopyranosiduronic acid (GUG-beta-CyD) (1) and its sodium salt were studied more extensively, as these compounds are most likely to have a practical application.