MilliporeSigma
Chemotactic penetration of keratocytes in ePTFE polymer in vitro.
Journal of biomedical materials research (2001-06-16)
F P Dupuy, M Savoldelli, A M Robert, L Robert, J M Legeais, G Renard
ABSTRACT
Expanded polytetrafluoroethylene (ePTFE) is used as a support for artificial corneas. Implanted in corneas, most of the time this polymer is colonized by corneal host cells. The absence of colonization often coincides with extrusion of the polymer. Therefore, we decided to introduce keratocytes into ePTFE in vitro before implantation. Because keratocytes do not spontaneously enter ePTFE, we used several chemoattractants, separately and in a mixture, to stimulate the penetration of cultured keratocytes into the polymer. The influence of the passage number on cell penetration was also studied. No significant differences were observed up to the seventh passage, although seventh-passage cells penetrated somewhat more slowly than younger cells. Satisfactory results were obtained with four of the tested chemotactic factors: IL-6, type alpha transforming growth factor (TGF-alpha), platelet derived growth factor isoform BB (PDGF-BB), and fibroblast growth factor-2 (FGF-2). Under our experimental conditions, two to more than six million keratocytes entered the polymer discs with a volume of 706.5 mm(3) in the presence of these four chemoattractants. TGF-alpha was the most efficient and was selected for further in vitro and in vivo studies.