MilliporeSigma
  • Gender differences in autoantibodies to oxidative DNA base damage in cigarette smokers.
Gender differences in autoantibodies to oxidative DNA base damage in cigarette smokers.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2001-06-13)
L A Mooney, F P Perera, A M Van Bennekum, W S Blaner, J Karkoszka, L Covey, Y Hsu, T B Cooper, K Frenkel
ABSTRACT
Oxidative DNA damage and antibodies to that damage have been implicated in lung, breast, and colorectal cancer. In this observational validation study, the relationship between anti-5-hydroxymethyl-2'-deoxyuridine (HMdU) autoantibody (aAb) and plasma micronutrients was assessed in 140 heavy smokers by ELISA. Anti-HMdU aAbs were 50% higher in women after adjustment for cigarettes/day (CPD; P = 0.002), although men smoked more and had higher plasma cotinine levels. The women reported taking more vitamin C (P < 0.005) and had higher plasma levels of alpha-carotene and beta-carotene (P < 0.001) and cryptoxanthin (P < 0.01) than men. Neither CPD nor cotinine was associated with aAb titers. Anti-HMdU aAbs were associated inversely with alpha-tocopherol (P = 0.10), retinol (P = 0.06), and age (P = 0.04) in women but not in men. In contrast to the men, women <or=50 years of age had significantly higher aAbs than those >50 years of age (P = 0.05). Given the same duration of exposure, women had higher anti-HMdU aAbs and also reached peak levels at a lower cumulative smoking exposure (30 years) compared with male smokers (40 years). Subjects smoked an average of 28.9 +/- 0.81 CPD and initiated smoking at 17.2 +/- 0.33 (SE) years of age. Therefore, smokers who reported smoking for 30 years were typically <50 years old. Women <or=50 years in the younger age group that smoked 21-30 years had significantly higher levels of aAbs than did men of the same age and smoking history (P = 0.012). Gender difference in aAbs was also evident in 29 persons who gave serial samples before and after quitting smoking (P < 0.028). In women, aAbs remained elevated for 14 months after smoking cessation but decreased significantly by 20.5 months (P < 0.032) by paired t tests. In men, aAbs increased with time since quitting smoking but not significantly. The finding of significantly elevated aAbs to oxidized DNA in females <or=50 compared with male smokers of the same age and exposure suggests a possible interaction with hormones (e.g., estrogens) and may explain a heightened risk of smoking-induced lung cancer in women compared with men.