MilliporeSigma
  • Insect growth inhibition, antifeedant and antifungal activity of compounds isolated/derived from Zingiber officinale Roscoe (ginger) rhizomes.
Insect growth inhibition, antifeedant and antifungal activity of compounds isolated/derived from Zingiber officinale Roscoe (ginger) rhizomes.
Pest management science (2001-07-18)
M Agarwal, S Walia, S Dhingra, B P Khambay
ABSTRACT
Fresh rhizomes of Zingiber officinale (ginger), when subjected to steam distillation, yielded ginger oil in which curcumene was found to be the major constituent. The thermally labile zingiberene-rich fraction was obtained from its diethyl ether extract. Column chromatography of ginger oleoresin furnished a fraction from which [6]-gingerol was obtained by preparative TLC. Naturally occurring [6]-dehydroshogaol was synthesised following condensation of dehydrozingerone with hexanal, whereas zingerone and 3-hydroxy-1-(4-hydroxy-3-methoxyphenyl)butane were obtained by hydrogenation of dehydrozingerone with 10% Pd/C. The structures of the compounds were established by 1H NMR, 13C NMR and mass (EI-MS and ES-MS) spectral analysis. The test compounds exhibited moderate insect growth regulatory (IGR) and antifeedant activity against Spilosoma obliqua, and significant antifungal activity against Rhizoctonia solani. Among the various compounds, [6]-dehydroshogaol exhibited maximum IGR activity (EC50 3.55 mg ml-1), while dehydrozingerone imparted maximum antifungal activity (EC50 86.49 mg litre-1).