MilliporeSigma
  • Secondary acute promyelocytic leukemia in a patient with non-Hodgkin's lymphoma treated with VP-16 and MST-16.
Secondary acute promyelocytic leukemia in a patient with non-Hodgkin's lymphoma treated with VP-16 and MST-16.
International journal of hematology (2002-02-15)
Takahiro Okamoto, Masaya Okada, Takeshi Wakae, Ako Mori, Hiroyuki Takatsuka, Eizo Kakishita