MilliporeSigma
  • Tyrosine kinase participates in vasoconstriction through a Ca(2+)- and myosin light chain phosphorylation-independent pathway.
Tyrosine kinase participates in vasoconstriction through a Ca(2+)- and myosin light chain phosphorylation-independent pathway.
FEBS letters (2002-02-20)
Lian Hua Fang, Seong Chun Kwon, Yong He Zhang, Hee Yul Ahn
ABSTRACT
This study was undertaken to determine the role of tyrosine kinase on intracellular Ca(2+) ([Ca(2+)](i)), myosin light chain (MLC) phosphorylation, and contraction caused by norepinephrine (NE) in rat aorta. NE induced a sustained contraction with an increase of [Ca(2+)](i). On the other hand, NE increased the phosphorylation of the 20 kDa MLC transiently. Pretreatment with genistein and tyrophostin 25, tyrosine kinase inhibitors, significantly inhibited NE-induced contraction, but did not affect the increase of [Ca(2+)](i) and MLC phosphorylation. These results suggest that tyrosine kinase may regulate the NE-mediated contraction without altering [Ca(2+)](i) and MLC phosphorylation in rat aorta.