MilliporeSigma
  • Certification of the critical importance of L-3-(2-naphthyl)alanine at position 3 of a specific CXCR4 inhibitor, T140, leads to an exploratory performance of its downsizing study.
Certification of the critical importance of L-3-(2-naphthyl)alanine at position 3 of a specific CXCR4 inhibitor, T140, leads to an exploratory performance of its downsizing study.
Bioorganic & medicinal chemistry (2002-03-12)
Hirokazu Tamamura, Akane Omagari, Kenichi Hiramatsu, Shinya Oishi, Hiromu Habashita, Taisei Kanamoto, Kazuyo Gotoh, Naoki Yamamoto, Hideki Nakashima, Akira Otaka, Nobutaka Fujii
ABSTRACT
We have previously found that a 14-amino acid residue-peptide, T140, inhibits infection of target cells by T cell line-tropic HIV-1 (X4-HIV-1) through its specific binding to a chemokine receptor, CXCR4. Here, the importance of an L-3-(2-naphthyl)alanine (Nal) residue at position 3 in T140 for high anti-HIV activity and inhibitory activity against Ca(2+) mobilization induced by stromal cell-derived factor (SDF)-1alpha-stimulation through CXCR4 has initially been shown by the synthesis and biological evaluation of several analogues, where Nal(3) is substituted by diverse aromatic amino acids. Next, the order of the N-terminal 3 residues (Arg(1)-Arg(2)-Nal(3)) has been proved to be important from the structure--activity relationship (SAR) study shuffling these residues. Based on these results, we have found 10-residue peptides possessing modest anti-HIV activity by systematic antiviral evaluation of a series of synthetic, shortened analogues of T140.