MilliporeSigma
  • Evaluation of 7-hydroxy-flavones as inhibitors of oestrone and oestradiol biosynthesis.
Evaluation of 7-hydroxy-flavones as inhibitors of oestrone and oestradiol biosynthesis.
Journal of enzyme inhibition (2002-03-28)
T K Vinh, P J Nicholls, A J Kirby, C Simons
ABSTRACT
A series of 4-aryl substituted 7-hydroxy-flavones were prepared using the three-step Baker-Venkataraman synthesis in good overall yields. The flavones were all evaluated in vitro for inhibitory activity against aromatase (P450AROM, CYP19), using human placental microsomes, and for inhibitory activity against 17beta-hydroxysteroid dehydrogenase type 1 (17beta-HSD-1) using human placental cytosol. The phenyl, 4-fluoro-phenyl and 4-bromo-phenyl derivatives displayed moderate inhibitory activity against P450AROM (IC50 17.2, 13.5 and 10.1 microM, respectively), none of the flavones, including the standard genistein, displayed any inhibitory activity against 17beta-HSD type 1 at 100 microM concentration.