MilliporeSigma
  • 7,8-Dihydroneopterin induces apoptosis of Jurkat T-lymphocytes via a Bcl-2-sensitive pathway.
7,8-Dihydroneopterin induces apoptosis of Jurkat T-lymphocytes via a Bcl-2-sensitive pathway.
European journal of cell biology (2002-05-23)
Christiane Enzinger, Barbara Wirleitner, Christina Lutz, Günther Böck, Bettina Tomaselli, Gottfried Baier, Dietmar Fuchs, Gabriele Baier-Bitterlich
ABSTRACT
Activated cell-mediated immunity is known to be accompanied by elevated concentrations of 7,8-dihydroneopterin which in high concentrations was found to interfere with the oxidant-antioxidant balance. In this study we investigated whether 7,8-dihydroneopterin mediates apoptosis of Jurkat T-lymphocytes via a CrmA- or Bcl-2-sensitive pathway. Transient transfection assays with CrmA and Bcl-2 expression constructs showed that apoptosis was not affected by CrmA whereas it was significantly decreased upon cotransfection with Bcl-2 constructs. Results suggest that 7,8-dihydroneopterin-induced apoptosis of T-lymphocytes is mediated by a Bcl-2-sensitive pathway.
Sigma-Aldrich
7,8-Dihydroneopterin, ≥97.0% (HPLC)