MilliporeSigma
  • 13C NMR signal detection of iron-bound cyanide ions in ferric cyanide complexes of heme proteins.
13C NMR signal detection of iron-bound cyanide ions in ferric cyanide complexes of heme proteins.
Journal of the American Chemical Society (2002-05-23)
Hiroshi Fujii
ABSTRACT
13CN ion appears to have the greatest potential to probe the heme environment of the ferric heme proteins; however, a resonance of the iron-bound (13)CN ion in ferric heme proteins has not yet been located. We show here the first detection of (13)C NMR signals of the iron-bound (13)CN for heme proteins and their model complexes in an unexpectedly large upfield region. This study demonstrates that the (13)C NMR signal of the iron-bound (13)CN is a sensitive probe to study the nature of the proximal ligand in ferric heme protein.
Sigma-Aldrich
Protoporphyrin IX dimethyl ester, ~90% (HPLC)