MilliporeSigma
  • Alverine citrate fails to relieve the symptoms of irritable bowel syndrome: results of a double-blind, randomized, placebo-controlled trial.
Alverine citrate fails to relieve the symptoms of irritable bowel syndrome: results of a double-blind, randomized, placebo-controlled trial.
Alimentary pharmacology & therapeutics (2002-05-29)
S A Mitchell, A S Mee, G D Smith, K R Palmer, R W Chapman
ABSTRACT
Alverine citrate has been used in the treatment of irritable bowel syndrome for many years. To compare the efficacy and safety of a new formulation of alverine citrate, a 120-mg capsule, with placebo given three times daily for 12 weeks. One hundred and seven patients with irritable bowel syndrome were entered into this three-centre, double-blind, randomized, placebo-controlled, parallel group trial. The primary end-point was relief of abdominal pain indicated by improvement in the scores for severity and frequency. Secondary efficacy variables included scores for other clinical symptoms and for overall well-being. The severity and frequency of abdominal pain improved in 66% and 68% of patients treated with alverine citrate vs. 58% and 69% of the placebo group, but these differences were not significant. The mean percentage reduction in the scores for abdominal pain from baseline to the final assessment, although greater in the alverine citrate group (43.7%) compared with the placebo group (33.3%), was not statistically significant. Alverine citrate is no better than placebo at relieving the symptoms of irritable bowel syndrome. Future trials should be designed to take into account the high and persistent placebo response seen in this condition.
Sigma-Aldrich
Alverine citrate salt
Pricing and availability is not currently available.
Alverine citrate salt, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.