MilliporeSigma
  • Syntheses of labeled vitamers of folic acid to be used as internal standards in stable isotope dilution assays.
Syntheses of labeled vitamers of folic acid to be used as internal standards in stable isotope dilution assays.
Journal of agricultural and food chemistry (2002-08-09)
Achim Freisleben, Peter Schieberle, Michael Rychlik
ABSTRACT
[2H4]Folic acid was synthesized by deuterating p-aminobenzoic acid, which was then coupled to glutamic acid and 6-formylpterin. Using [2H4]folic acid as starting component enabled the preparation of labeled vitamers tetrahydrofolate, 5-formyltetrahydrofolate, 5-methyltetrahydrofolate, and 10-formylfolate which were characterized by electrospray mass spectrometry and collision-induced dissociation. The mass spectrometric studies confirmed that the compounds could be used as internal standards in stable isotope dilution assays.