MilliporeSigma
  • Tracing the source of 3,3'-dichlorobiphenyl found in samples collected in and around Halifax Harbour.
Tracing the source of 3,3'-dichlorobiphenyl found in samples collected in and around Halifax Harbour.
Marine pollution bulletin (2002-09-12)
T L King, P Yeats, J Hellou, S Niven
ABSTRACT
3,3'-dichlorobiphenyl (IUPAC No. 11), a chlorobiphenyl (CB) that is not generally analysed in environmental studies of CBs, is found, sometimes at high concentrations, in water, suspended particulate material, biota and sediments from Halifax Harbour, NS, Canada. The results presented demonstrate the need for investigations of non-Aroclor CBs like CB 11. Not only can they have rather elevated environmental concentrations like those reported here for CB 11, but they also can be members of the more toxic non-ortho class of CBs and thus important from a toxicity standpoint. The focus of this paper was to investigate the possible sources of CB 11, a trace constituent of commercial mixtures, but dominant in various environmental compartments of Halifax Harbour.