MilliporeSigma
  • Polycyclic aromatic hydrocarbon o-quinones inhibit the activity of the catalytic fragment of protein kinase C.
Polycyclic aromatic hydrocarbon o-quinones inhibit the activity of the catalytic fragment of protein kinase C.
Biochemistry (2002-09-25)
Deshan Yu, Marcelo G Kazanietz, Ronald G Harvey, Trevor M Penning
ABSTRACT
Polycyclic aromatic hydrocarbons (PAHs) require metabolic activation to exert their carcinogenic effects. PAH trans-dihydrodiol proximate carcinogens are oxidized by aldo-keto reductases (AKRs) to their corresponding reactive and redox-active o-quinones which may have the properties of initiators and promoters. To determine whether these o-quinones target protein kinase C (PKC), their effects on human recombinant PKCalpha and PKCdelta and the catalytic fragment of rat brain PKC were determined. Naphthalene-1,2-dione (NP-1,2-dione), benzo[a]pyrene-7,8-dione (BP-7,8-dione), and 7,12-dimethylbenz[a]anthracene-3,4-dione (DMBA-3,4-dione) potently inhibited (IC(50) values 3-5 microM) the basal and stimulated activity of the holoenzymes PKCalpha and PKCdelta in a dose-dependent manner. Inhibition of PKC by BP-7,8-dione was observed irrespective of whether PKCalpha activity was stimulated with phorbol 12-myristate 13-acetate (PMA), phosphatidylserine (PS), or Ca(2+) or whether PKCdelta was stimulated with phorbol 12-myristate 13-acetate (PMA) or phosphatidylserine (PS), suggesting that the inhibition was not cofactor-specific. All three quinones inhibited the catalytic fragment of PKC in vitro, yielding identical IC(50) values (3-5 microM), indicating that they interact with the catalytic domain of PKC rather than the cofactor/activator sites. In contrast, no effect on either the holoenzyme or the catalytic fragment was observed with the corresponding PAH trans-dihydrodiols, indicating that inhibition was o-quinone-specific. Irreversible inhibition of the catalytic fragment of PKC was observed since activity could not be restored by dialysis, suggesting that arylation of the fragment had occurred. NP-1,2-dione and BP-7,8-dione also suppressed PKC activity in human breast cancer MCF-7 cell lysates which express PKCalpha, -beta, -delta, -epsilon, -iota, and -lambda isozymes. These data suggest that PAH o-quinones, generated by AKRs, may affect cellular signaling through suppression of the activity of PKC isoforms.