MilliporeSigma
  • Prodrugs to enhance central nervous system effects of the TRH-like peptide pGlu-Glu-Pro-NH2.
Prodrugs to enhance central nervous system effects of the TRH-like peptide pGlu-Glu-Pro-NH2.
Bioorganic & medicinal chemistry letters (2003-03-20)
Katalin Prokai-Tatrai, Vien Nguyen, Alevtina D Zharikova, April C Braddy, Stanley M Stevens, Laszlo Prokai
ABSTRACT
Potential prodrugs for the TRH-like tripeptide pGlu-Glu-Pro-NH(2) were synthesized either by esterifying the Glu side-chain of the parent peptide in solution with alcohols in the presence of resin-bound dicyclohexylcarbodiimide or by solid-phase peptide chemistry. Affinities of these ester prodrugs to lipid membranes as predictors of the transport across the blood-brain barrier were compared by immobilized artificial membrane chromatography, and prodrug activation was tested in the brain tissue of experimental animals. Esters of pGlu-Glu-Pro-NH(2) with long-chain primary alcohols emerged as potentially useful prodrugs to improve the central nervous system activity of pGlu-Glu-Pro-NH(2) upon systemic administration, as revealed by the enhancement of analeptic activity in mice.