MilliporeSigma
  • 166Ho-DOTMP plus melphalan followed by peripheral blood stem cell transplantation in patients with multiple myeloma: results of two phase 1/2 trials.
166Ho-DOTMP plus melphalan followed by peripheral blood stem cell transplantation in patients with multiple myeloma: results of two phase 1/2 trials.
Blood (2003-05-06)
Sergio Giralt, William Bensinger, Mark Goodman, Donald Podoloff, Janet Eary, Richard Wendt, Raymond Alexanian, Donna Weber, David Maloney, Leona Holmberg, Joseph Rajandran, Hazel Breitz, Richard Ghalie, Richard Champlin
ABSTRACT
Holmium-166 1, 4, 7, 10-tetraazcyclododecane-1, 4, 7, 10-tetramethylenephosphonate (166Ho-DOTMP) is a radiotherapeutic that localizes specifically to the skeleton and can deliver high-dose radiation to the bone and bone marrow. In patients with multiple myeloma undergoing autologous hematopoietic stem cell transplantation two phase 1/2 dose-escalation studies of high-dose 166Ho-DOTMP plus melphalan were conducted. Patients received a 30 mCi (1.110 Gbq) tracer dose of 166Ho-DOTMP to assess skeletal uptake and to calculate a patient-specific therapeutic dose to deliver a nominal radiation dose of 20, 30, or 40 Gy to the bone marrow. A total of 83 patients received a therapeutic dose of 166Ho-DOTMP followed by autologous hematopoietic stem cell transplantation 6 to 10 days later. Of the patients, 81 had rapid and sustained hematologic recovery, and 2 died from infection before day 60. No grades 3 to 4 nonhematologic toxicities were reported within the first 60 days. There were 27 patients who experienced grades 2 to 3 hemorrhagic cystitis, only 1 of whom had received continuous bladder irrigation. There were 7 patients who experienced complications considered to be caused by severe thrombotic microangiopathy (TMA). No cases of severe TMA were reported in patients receiving in 166Ho-DOMTP doses lower than 30 Gy. Approximately 30% of patients experienced grades 2 to 4 renal toxicity, usually at doses targeting more than 40 Gy to the bone marrow. Complete remission was achieved in 29 (35%) of evaluable patients. With a minimum follow-up of 23 months, the median survival had not been reached and the median event-free survival was 22 months. 166Ho-DOTMP is a promising therapy for patients with multiple myeloma and merits further evaluation.