MilliporeSigma
  • Synthesis of S6-(2,4-dinitrophenyl)-6-thioguanosine phosphoramidite and its incorporation into oligoribonucleotides.
Synthesis of S6-(2,4-dinitrophenyl)-6-thioguanosine phosphoramidite and its incorporation into oligoribonucleotides.
Bioorganic & medicinal chemistry letters (2003-09-03)
Qinguo Zheng, Yang Wang, Eric Lattmann
ABSTRACT
The preparation of N(2)-phenoxylacetyl-S(6)-(2,4-dinitrophenyl)-6-thioguanosine phosphoramidite and its subsequent incorporation into oligoribonucleotides is described. The identity of the oligonucleotides was confirmed by UV spectrophotometry and nucleoside composition analysis.