MilliporeSigma
SURGICAL MANAGEMENT OF REGIONAL ENTERITIS.
Canadian Medical Association journal (1964-07-04)
M M LASKIN
ABSTRACT
The present-day concepts concerning the surgical management of regional enteritis are reviewed and the multitude of problems that may arise in surgical treatment are discussed. The primary treatment of regional enteritis remains medical. Surgical intervention is necessary only for the complications of intractability, obstruction, fistula, abscess formation, anal and rectal complications, massive hemorrhage and perforation. To ensure the best possible results, medical treatment should continue after surgery. A perfect operation for this disease does not exist. Operations for regional enteritis can now be performed with a mortality rate of less than 2%, and although the recurrence rate following surgery averages approximately 30%, the disease is well controlled in the majority of patients with medical and/or surgical treatment.