MilliporeSigma
  • Action of prostanoids on the emetic reflex of Suncus murinus (the house musk shrew).
Action of prostanoids on the emetic reflex of Suncus murinus (the house musk shrew).
European journal of pharmacology (2003-10-03)
Kelvin K W Kan, Robert L Jones, Man P Ngan, John A Rudd
ABSTRACT
Several prostanoids were investigated for a potential to induce emesis in Suncus murinus. The TP receptor agonist 11alpha,9alpha-epoxymethano-15S-hydroxyprosta-5Z,13E-dienoic acid (U46619) induced emesis at doses as low as 3 microg/kg, i.p. but the DP receptor agonist 5-(6-Carboxyhexyl)-1-(3-cyclohexyl-3-hydroxypropyl) hydantoin (BW245C) was approximately 1000 times less potent. The emetic action of U46619 (300 microg/kg, i.p.) was antagonized significantly by the TP receptor antagonist, vapiprost (P<0.05). EP (prostaglandin E(2), 17-phenyl-omega-trinor prostaglandin E(2), misoprostol and sulprostone), FP (prostaglandin F(2alpha) and fluprostenol) and IP (iloprost and cicaprost) receptor agonists failed to induce consistent emesis at doses up to 300-1000 microg/kg, i.p. Fluprostenol reduced nicotine (5 mg/kg, s.c.)-but not copper sulphate (120 mg/kg, intragastric)-induced emesis; the other inconsistently emetic prostanoids were inactive to modify drug-induced emesis. The results indicate an involvement of TP and possibly DP and FP receptors in the emetic reflex of S. murinus.