MilliporeSigma
  • Glucocorticoids modulate neurotransmitter-induced glycogen metabolism in cultured cortical astrocytes.
Glucocorticoids modulate neurotransmitter-induced glycogen metabolism in cultured cortical astrocytes.
Journal of neurochemistry (2004-02-06)
Igor Allaman, Luc Pellerin, Pierre J Magistretti
ABSTRACT
Glucocorticoids (GC) are considered as key modulators of glycogen homeostasis in peripheral tissues, but their role in the central nervous system has only partially been characterized. Exposure of primary cultures of cortical astrocytes to dexamethasone (DEX), a synthetic glucocorticoid, results in the reduction of noradrenaline (NA)-induced glycogen synthesis in a concentration-dependent manner with a IC50 of 4.88 nm and a maximum inhibition of 51%. Such an effect is mediated via glucocorticoid receptors (GRs), since it is mimicked by the glucocorticoid analogue RU28362 (100 nm) and prevented by the GR antagonist RU38486 (1 micro m). DEX does not act through alteration of signal transduction mechanisms, as cAMP formation induced by noradrenergic stimulation was unchanged. Moreover, glycogen synthesis was inhibited to the same extent when DEX was applied either together or only after a brief NA application. Neither [3H]2-deoxyglucose uptake nor lactate release was altered by DEX in the presence of NA, demonstrating that inhibition of glycogen synthesis is not a consequence of reduced glucose utilization or availability. Interestingly, enhancement of glycogen synthase activity induced by NA was reduced in the presence of DEX (-27%). These results suggest that GC could have a significant influence on neuroenergetics as they could modulate activity-related changes in brain glycogen metabolism.