MilliporeSigma
  • Estrogen receptor enhances the antiproliferative effects of trichostatin A and HC-toxin in human breast cancer cells.
Estrogen receptor enhances the antiproliferative effects of trichostatin A and HC-toxin in human breast cancer cells.
Archives of pharmacal research (2004-06-19)
Kyung Nan Min, Min Jung Cho, Dae-Kee Kim, Yhun Yhong Sheen
ABSTRACT
Trichostatin A, an antifungal antibiotics, and HC-toxin are potent and specific inhibitors of histone deacetylase activity. Histone deacetylase inhibitors are new class of chemotherapeutic drugs able to induce tumor cell apoptosis and/or cell cycle arrest. In this study, the antiproliferative activities of trichostatin A and HC-toxin were compared between estrogen receptor positive human breast cancer cell MCF-7 and estrogen receptor negative human breast cancer cell MDA-MB-468. Trichostatin A and HC-toxin showed potent antiproliferative activity in both MCF-7 and MDA-MB-468 cells. In MCF-7 cells that contain high level estrogen receptor, trichostatin A and HC-toxin brought about three-times more potent cell growth inhibitory effect than estrogen receptor negative MDA-MB-468 cells. Both trichostatin A and HC-toxin showed cell cycle arrest at G2/M phases of MCF-7 and MDA-MB-468 cells in a dose- and time-dependent manner. Trichostatin A and HC-toxin also induced apoptosis from MCF-7 and MDA-MB-468 cells in a dose- and time-dependent manner. Results of this study suggested that antiproliferative effects of trichostatin A and HC-toxin might be involved in estrogen receptor signaling pathway, but cell cycle arrest and apoptosis of trichostatin A and HC-toxin might not be involved in estrogen receptor system of human breast cancer cells.