MilliporeSigma
  • Repeatedly usable immobilized pH gradient in a monolithic capillary column.
Repeatedly usable immobilized pH gradient in a monolithic capillary column.
Electrophoresis (2004-06-24)
Chun Yang, Guijie Zhu, Lihua Zhang, Weibing Zhang, Yukui Zhang
ABSTRACT
Glycidyl methacrylate (GMA) and ethylene dimethacrylate (EDMA) were used to synthesize a monolithic capillary column containing reactive epoxy groups. Glutaraldehyde was introduced and linked to the monolith after a process of amination. An aqueous solution of commercial carrier ampholytes (CAs, Ampholine) was focused in such a polymer column. The primary amino groups of CAs reacted with glutaraldehyde along the capillary. CAs were immobilized at different positions in the column according to their isoelectric points (pI), resulting in a monolithic immobilized pH gradient (M-IPG). Isoelectric focusing (IEF) was performed without CAs in such an M-IPG column. Due to the covalent attachment of the CAs this M-IPG can be repeatedly used after its preparation. Good stability, linearity, and reproducibility were obtained.