MilliporeSigma
  • First total synthesis of the antitumor compound (-)-kazusamycin A and absolute structure determination.
First total synthesis of the antitumor compound (-)-kazusamycin A and absolute structure determination.
Organic letters (2004-08-28)
Noriyoshi Arai, Noriko Chikaraishi, Satoshi Omura, Isao Kuwajima
ABSTRACT
The first total synthesis of kazusamycin A, a potent antitumor compound from an actinomycete, has been achieved, and its absolute structure was determined. Paterson's stereoselective aldol reaction was successfully applied to construct the contiguous chiral centers by using an originally designed optically active 2-acyl-1,3-propanediol derivative.