MilliporeSigma
Metabolic fate of orally administered phyllodulcin in rats.
Journal of natural products (2004-09-25)
Takaaki Yasuda, Saiko Kayaba, Kana Takahashi, Takahiro Nakazawa, Keisuke Ohsawa
ABSTRACT
Naturally occurring phyllodulcin (1) was orally administered to rats to investigate its metabolic fate. Urinary metabolites were analyzed by three-dimensional HPLC. Phyllodulcin-3'-O-sulfate (2), phyllodulcin-3'-O-beta-glucuronide (3), 2-[2-(3,4-dihydroxyphenyl)ethyl]-6-hydroxybenzoic acid (4), and one novel bibenzyl derivative, 2-[2-(3-hydroxy-4-methoxyphenyl)ethyl]-6-hydroxybenzoic acid (5), together with thunberginol G (6) and hydrangenol (7) were isolated from the phyllodulcin-treated urine. 1 was extensively metabolized to 4-6 by a rat fecal suspension after incubation for 24 h. Urinary excretion of 4-6 in rats administered phyllodulcin orally was substantially reduced when the rats were treated with antibiotics to suppress their intestinal flora. On the other hand, the incubation of 1 with rat liver S-9 mix showed the presence of 7 together with 4 and 5.