MilliporeSigma
  • Bidirectional regulation of renal cortical Na+,K+-ATPase by protein kinase C.
Bidirectional regulation of renal cortical Na+,K+-ATPase by protein kinase C.
Acta biochimica Polonica (2004-09-28)
Jerzy Bełtowski, Andrzej Marciniak, Anna Jamroz-Wiśniewska, Ewelina Borkowska, Grazyna Wójcicka
ABSTRACT
We examined the role of protein kinase C (PKC) in the regulation of Na+,K+- ATPase activity in the renal cortex. Male Wistar rats were anaesthetized and the investigated reagents were infused into the abdominal aorta proximally to the renal arteries. A PKC-activating phorbol ester, phorbol 12,13-dibutyrate (PDBu), had a dose-dependent effect on cortical Na+,K+-ATPase activity. Low dose of PDBu (10(-11) mol/kg per min) increased cortical Na+,K+-ATPase activity by 34.2%, whereas high doses (10(-9) and 10(-8) mol/kg per min) reduced this activity by 22.7% and 35.0%, respectively. PDBu administration caused changes in Na+,K+-ATPase Vmax without affecting K(0.5) for Na+, K+ and ATP as well as Ki for ouabain. The effects of PDBu were abolished by PKC inhibitors, staurosporine, GF109203X, and Gö 6976. The inhibitory effect of PDBu was reversed by pretreatment with inhibitors of cytochrome P450-dependent arachidonate metabolism, ethoxyresorufin and 17-octadecynoic acid, inhibitors of phosphatidylinositol 3-kinase (PI3K), wortmannin and LY294002, and by actin depolymerizing agents, cytochalasin D and latrunculin B. These results suggest that PKC may either stimulate or inhibit renal cortical Na+,K+-ATPase. The inhibitory effect is mediated by cytochrome P450-dependent arachidonate metabolites and PI3K, and is caused by redistribution of the sodium pump from the plasma membrane to the inactive intracellular pool.