MilliporeSigma
  • Etiological and therapeutical observations in a case of belly dancer's dyskinesia.
Etiological and therapeutical observations in a case of belly dancer's dyskinesia.
Movement disorders : official journal of the Movement Disorder Society (2004-09-30)
Girutz Linazasoro, Nadège Van Blercom, Asier Lasa, José Manuel Fernández, Inés Aranzábal
ABSTRACT
We report on the case of a woman with belly dancer's syndrome. This case presented two peculiarities: (1) the condition was induced by the chronic use of clebopride, and (2) abdominal dyskinesias showed a dramatic response to the application of transcutaneous electrical nerve stimulation.