MilliporeSigma
  • Crystal structures of calpain-E64 and -leupeptin inhibitor complexes reveal mobile loops gating the active site.
Crystal structures of calpain-E64 and -leupeptin inhibitor complexes reveal mobile loops gating the active site.
Journal of molecular biology (2004-10-20)
T Moldoveanu, R L Campbell, D Cuerrier, P L Davies
ABSTRACT
The endogenous calpain inhibitor, calpastatin, modulates some patho-physiological aspects of calpain signaling. Excess calpain can escape this inhibition and as well, many calpain isoforms and autolytically generated protease core fragments are not inhibited by calpastatin. There is a need, therefore, to develop specific, cell-permeable calpain inhibitors to block uncontrolled proteolysis and prevent tissue damage during brain and heart ischemia, spinal-cord injury and Alzheimer's diseases. Here, we report the first high-resolution crystal structures of rat mu-calpain protease core complexed with two traditional, low molecular mass inhibitors, leupeptin and E64. These structures show that access to a slightly deeper, but otherwise papain-like active site is gated by two flexible loops. These loops are divergent among the calpain isoforms giving a potential structural basis for substrate/inhibitor selectivity over other papain-like cysteine proteases and between members of the calpain family.