MilliporeSigma
  • Retinoic acid induces neuroblastoma cell death by inhibiting proteasomal degradation of retinoic acid receptor alpha.
Retinoic acid induces neuroblastoma cell death by inhibiting proteasomal degradation of retinoic acid receptor alpha.
Cancer research (2004-11-03)
Jun-ichi Nagai, Takuya Yazawa, Koji Okudela, Hisato Kigasawa, Hitoshi Kitamura, Hitoshi Osaka
ABSTRACT
To seek a novel therapeutic approach to neuroblastoma (NBL), we used three NBL cell lines (SK-N-DZ, NH12, and SK-N-SH) to examine the underlining molecular mechanisms of cellular reactions and sensitivity to all-trans-retinoic acid (ATRA). SK-N-DZ cells expressed relatively high levels of retinoic acid receptor alpha (RAR-alpha) and underwent ATRA-induced cell death that was blocked by an RAR-alpha antagonist. By contrast, RAR-alpha expression gradually decreased in NH12 and SK-N-SH cells, which did not experience increased cell death in response to ATRA. We report here the ubiquitin-dependent down-regulation of RAR-alpha expression during ATRA treatment. Our data suggest that SK-N-DZ cells have a defect in RAR-alpha down-regulation, resulting in sustained high expression of RAR-alpha that confers high sensitivity to ATRA. Accordingly, treatment with a proteasome inhibitor dramatically increased ATRA-induced cell death in NH12 and SK-N-SH cell lines. Our results reveal the crucial involvement of the RAR-alpha signaling pathway in NBL cell death and show that three NBL cell lines are differentially sensitive to ATRA. These data suggest a potential novel therapy for NBL involving retinoic acid treatment combined with the inhibition of RAR-alpha degradation.
Sigma-Aldrich
Ro 41-5253, ≥98% (HPLC)