MilliporeSigma
  • Calculation of the peak concentration of 2-chlorobenzamide generated in the hydrolysis of the benzoylphenylurea insecticide 1-(2-chlorobenzoyl)-3-(4-chlorophenyl)urea.
Calculation of the peak concentration of 2-chlorobenzamide generated in the hydrolysis of the benzoylphenylurea insecticide 1-(2-chlorobenzoyl)-3-(4-chlorophenyl)urea.
Journal of agricultural and food chemistry (2004-12-23)
Wenying Lu, Qingxiang Zhou, Guoguang Liu
ABSTRACT
1-(2-chlorobenzoyl)-3-(4-chlorophenyl)urea (CCU), a new analogue of diflubenuron and PH-6038, has been widely used in agriculture and forestry as a molt-inhibiting hormone insecticide which was developed in China. 2-Chlorobenzamide, a main degradation product of CCU in the environment, has been identified as a potential carcinogen, so the content of 2-chlorobenzamide from the breakup of CCU will directly affect the environmental safety of CCU. In this paper we describe a simple, rapid, and convenient prediction model for predicting the level and time of occurrence of the peak concentration of 2-chlorobenzamide in the hydrolysis of CCU verified by experimental data. The time for reaching the peak concentration of 2-chlorobenzamide (tm) at 25 degrees C and pH 6 is 13.5 d, and the maximum concentration of 2-chlorobenzamide (ym) is 3.2% of the initial concentration of CCU according to the results from the prediction model. These results are similar to the real values from the experiments, which are 22 d and 1.6% of the initial concentration of CCU, respectively. The difference between the values of the prediction and experiment is discussed, and it is demonstrated that the predicting model is highly credible.