MilliporeSigma
  • Accuracy of computed tomographic intravenous cholangiography (CT-IVC) with iotroxate in the detection of choledocholithiasis.
Accuracy of computed tomographic intravenous cholangiography (CT-IVC) with iotroxate in the detection of choledocholithiasis.
European radiology (2005-01-01)
Robert N Gibson, Janette M Vincent, Tony Speer, Neil A Collier, Keith Noack
ABSTRACT
To determine the accuracy of computed tomographic intravenous cholangiography (CT-IVC) for detection of choledocholithiasis. Sixty-five patients undergoing endoscopic retrograde cholangiography (ERC) underwent CT-IVC prior to ERC, using a single detector helical CT following intravenous infusion of 100 ml iotroxate. Patients with bilirubin levels >3 times normal were excluded. ERC was indeterminate in three patients (4.7%) and CT-IVC in four (6.3%). Twenty-three patients had ductal calculi at ERC, and CT-IVC was positive in 22, with two false positives and one false negative: sensitivity 95.6%, specificity 94.3%. Stones were multiple in nine and solitary in 14. Of the 14 solitary stones, ten were <or=5 mm and eight were <or=4 mm. The bilirubin level in positive cases was within normal in 20. Maximum intensity projection (MIP) reformats showed stones in only 27% of cases and surface rendered (SR) reformats in none. CT-IVC is highly accurate for detection of ductal calculi, including single small calculi, with a normal or near normal serum bilirubin. Axial images should be used for interpretation rather than MIP or SR reformats.