MilliporeSigma
  • Inhibition of the uptake of oxidized low-density lipoprotein in macrophage J774 by the antibiotic ikarugamycin.
Inhibition of the uptake of oxidized low-density lipoprotein in macrophage J774 by the antibiotic ikarugamycin.
European journal of biochemistry (1992-04-15)
K Hasumi, C Shinohara, S Naganuma, A Endo
ABSTRACT
The antiprotozoal antibiotic ikarugamycin was found to significantly inhibit oxidized low-density lipoprotein(LDL)-induced accumulation of cholesteryl ester in macrophage J774 at a concentration over 1-4 microM. Cholesteryl ester synthesis from [14C]oleate in the macrophages was similarly inhibited by the antibiotic, while the synthesis of triacylglycerol and polar lipids was not affected. The internalization of oxidized [125I]LDL in macrophages was reduced to 50% by 2 microM ikarugamycin, while cell-surface binding of oxidized [125I]LDL, lysosomal hydrolysis of the internalized oxidized [125I]LDL and microsomal acyl-coenzyme A:cholesterol acyltransferase was not detectably inhibited by 5 microM ikarugamycin. The results demonstrated that ikarugamycin inhibited cholesteryl ester accumulation in macrophage J774 by specifically inhibiting the uptake of oxidized LDL.